Pliers, Replaceable Tip, Straight, Excelta
Pliers, Replaceable Tip, Straight, Excelta
Supplier:   Excelta
Pliers, Replaceable Tip, Straight, Excelta
Supplier:  Excelta
Certificates
Pliers, Replaceable Tip, Straight, Excelta
Pliers, Replaceable Tip, Straight, Excelta
7 Options Available Below
Product Details & Documents
These pliers are useful in straightening leads and wires when metal-to-metal contact is not desired.
  • Anti-shock trimming of leads to specific length
  • Ergonomic grips for comfort
  • Double leaf springs for ease of use
This series of pliers offer soft jaw inserts to protect delicate parts. The jaw inserts are factory replaceable, and offered in Acetal, Brass and Nylon. The overall length is 5.75". They feature an ergonomically molded antimicrobial grip (10¹⁰ ohms/sq surface resistivity) with dual leaf springs.
Product Family Options
Acetal Jaws
Product Information
  • Désignation
    Matière
    Longueur
    Tip Size
    Curvature
    Availability
    Price
  • Pliers, replaceable tip, 5 star quality
    Acetal
    143.75 mm (5.75")
    3.75×3.75 mm (0.15×0.15")
    Straight
    Log in for availability
    Log in for availability
    Each
    Each - $503.61
    $503.61
    Each
       $503.61
    Supplier Number: 531A-US
    Specifications:
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    63042-552
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
  • Pliers, replaceable tip, 5 star quality
    Acetal
    143.75 mm (5.75")
    2×2 mm (0.08×0.08")
    Straight
    Log in for availability
    Log in for availability
    Each
    Each - $548.72
    $548.72
    Each
       $548.72
    Supplier Number: 532-US
    Specifications:
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    63042-560
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
Brass Jaws
Product Information
  • Désignation
    Matière
    Longueur
    Tip Size
    Curvature
    Availability
    Price
  • Pliers, replaceable tip, 5 star quality
    Brass
    143.75 mm (5.75")
    3.75×3.75 mm (0.15×0.15")
    Straight
    Log in for availability
    Log in for availability
    Each
    Each - $505.49
    $505.49
    Each
       $505.49
    Supplier Number: 531B-US
    Specifications:
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    63042-554
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
  • Pliers, replaceable tip, 5 star quality
    Brass
    143.75 mm (5.75")
    2×2 mm (0.08×0.08")
    Straight
    Log in for availability
    Log in for availability
    Each
    Each - $561.48
    $561.48
    Each
       $561.48
    Supplier Number: 532B-US
    Specifications:
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    63042-558
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
Nylon Jaws
Product Information
  • Désignation
    Matière
    Longueur
    Tip Size
    Curvature
    Availability
    Price
  • Pliers, flat/round, 1 star quality
    Stainless steel/nylon inserts
    137.5 mm (5.5")
    Straight
    Discontinued
    Supplier Number: -
    Specifications:
    More
    Specifications
    Cat. No.
    75880-934
  • Pliers, replaceable tip, 1 star quality
    Stainless steel/nylon inserts
    137.5 mm (5.5")
    Straight
    Log in for availability
    Log in for availability
    Each
    Each - $79.84
    $79.84
    Each
       $79.84
    Supplier Number: 2831
    Specifications:
    More
    Specifications
    Cat. No.
    89411-774
  • Pliers, replaceable tip, 5 star quality
    Nylon
    143.75 mm (5.75")
    3.75×3.75 mm (0.15×0.15")
    Straight
    Log in for availability
    Log in for availability
    Each
    Each - $538.75
    $538.75
    Each
       $538.75
    Supplier Number: 531-US
    Specifications:
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    63042-556
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
PFTE Jaws
Product Information
  • Désignation
    Matière
    Longueur
    Tip Size
    Curvature
    Availability
    Price
  • Pliers, replaceable tip, 5 star quality
    PTFE
    150 mm (6")
    Straight
    Log in for availability
    Log in for availability
    Each
    Each - $550.88
    $550.88
    Each
       $550.88
    Supplier Number: 533-US
    Specifications:
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    63042-564
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise