Wire Stripper, Excelta
 :   Excelta
Wire Stripper, Excelta
 :  Excelta
Wire Stripper, Excelta
Wire Stripper, Excelta
These two tools are designed for stripping PTFE coated small wires.
  • Will strip PTFE coating in one shot
  • Ergonomic grips for comfort
  • Double leaf springs for ease of use
These tools are made to strip small PTFE coated wire up to 30AWG. The 502-US will strip one size wire; (specify the wire gauge size by indicating the gauge at the end of the reference number. Available in 22, 24, 26, 28, and 30 AWG.) The 510E-US is a 'V' notched wire stripper which cuts the insulation on four sizes makeing stripping much easier. It features a ergonomically molded antimicrobial grip (10¹⁰ ohms/sq surface resistivity) with dual leaf springs. Both are made of carbon tool steel with flame hardened edges.
Product Family Options
  • Désignation
    Matière
    Longueur
  • Pliers, wire stripper, 5 star quality, custom size wire
    Carbon
    137.5 mm (5.5")
     : 502E-US
  • Pliers, wire stripper, v-notch, 5 star quality, adjustable
    Carbon
    137.5 mm (5.5")
    Each
    Each - $810.75
    $810.75
    Each
       $810.75
     : 510E-US