Pliers, Concave/Convex, Excelta
Pliers, Concave/Convex, Excelta
Supplier:   Excelta
Pliers, Concave/Convex, Excelta
Supplier:  Excelta
Certificates
Pliers, Concave/Convex, Excelta
Pliers, Concave/Convex, Excelta
2 Options Available Below
Product Details & Documents
These are designed for forming a 'J Hook' on wires and leads.
  • Specifically designed for forming 'J hook'
  • Ergonomic grips for comfort
  • Double leaf springs for ease of use
These small convex/concave forming plier are designed for forming a 'J Hook' on wires and leads. The jaw tapers from front to back for forming various lead diameters. They feature an ergonomically molded antimicrobial grip (10¹⁰ ohms/sq surface resistivity) with dual leaf springs. They are made of carbon tool steel.
Product Family Options
Product Information
  • Désignation
    Matière
    Longueur
    Availability
    Price
  • Pliers, concave/convex, 5 star quality, 0.12" tip height
    Carbon
    131.25 mm (5.25")
    Log in for availability
    Log in for availability
    Each
    Each - $1,046.75
    $1,046.75
    Each
       $1,046.75
    Supplier Number: 500-101-US
  • Pliers, concave/convex, 5 star quality, 0.1" tip height
    Carbon
    137.5 mm (5.5")
    Log in for availability
    Log in for availability
    Each
    Each - $1,044.66
    $1,044.66
    Each
       $1,044.66
    Supplier Number: 500-101A-US