VWR® Disposable Face Shields
Disposable face shields, 0.25 mm
Perfect for use in environments with light particle, splash, or spray, the VWR® Disposable Face Shield is a cost effective and reliable way to keep covered.
- Material: APET
- Anti-fog inner and outer coating
- PE protective film
- Comfortable PU foam for extended wear
- Elastic band to maintain comfortable fit
Face shields should be worn with ANSI approved respiratory protection, as needed.
: Packaged 200 per case.
- Désignation:Disposable face shields
- Épaisseur:0.25 mm