Brady® BradyPrinter i7100 Vial Label Applicator, Brady Worldwide
Catalogue # 76461-900
Fournisseur: Brady Worldwide
Spécifications
- For Use With:BradyPrinter i7100 Industrial Label Printer
- Description:Vial label applicator
- Cat. no.:76461-900
- Dimensions:4.75H×3W×3" D
- Label size:2.2" max. label/tape width
Spécifications
À propos de cet article
Automate your vial identification with the BradyPrinter i7100 Vial Label Applicator. This accessory allows you to keep high-quality data during your high-volume vial identification projects.
Work with your BradyPrinter i7100 Industrial Label Printer peel models by applying your labels on to the vial as they print. Pays for itself - after 100 hours of use, this accessory will save you more time and money that handwriting and hand-applying labels. Saves your lab techs both time and money by reducing labeling time. Works only with the 300 and 600 dpi Peel models of the BradyPrinter i7100 Industrial Label Printers.