SmartWall® Uprights
SmartWall® Uprights
Supplier:   Metropolitan Wire
SmartWall® Uprights
Supplier:  Metropolitan Wire
Certificates
SmartWall® Uprights
SmartWall® Uprights
5 Options Available Below
Product Details & Documents
Create a unique storage and workstation system for underutilized wall space with SmartWall® Uprights.
  • Built-in slots allow shelf supports and grids to attach without tools or hardware
  • 1.5" (38 mm) slot spacing allows shelves and grids to be adjusted on 1.5" (38 mm) increments
  • Designed for medium-duty applications
  • Two uprights are required for a stand-alone unit
  • Each part number is one individual upright
Corrosion-resistant Metroseal Gray epoxy finish includes Microban® antimicrobial product protections to continuously protect the product and keep it cleaner in between cleanings.
Product Family Options
Product Information
  • Désignation
    Color
    Finish
    Matière
    Largeur
    Profondeur
    Hauteur
    Availability
    Price
  • Upright
    Grey
    Metroseal epoxy
    Steel
    1⅛"
    1⅞"
    16"
    Log in for availability
    Log in for availability
    Each
    Each - $139.00
    $139.00
    Each
       $139.00
    Minimum order quantity is 1
    Supplier Number: SWU15K4
  • Upright
    Grey
    Metroseal epoxy
    Steel
    1⅛"
    1⅞"
    31"
    Log in for availability
    Log in for availability
    Each
    Each - $164.00
    $164.00
    Each
       $164.00
    Minimum order quantity is 1
    Supplier Number: SWU30K4
  • Upright
    Grey
    Metroseal epoxy
    Steel
    1⅛"
    1⅞"
    44½"
    Log in for availability
    Log in for availability
    Each
    Each - $189.00
    $189.00
    Each
       $189.00
    Minimum order quantity is 1
    Supplier Number: SWU45K4
  • Upright
    Grey
    Metroseal epoxy
    Steel
    1⅛"
    1⅞"
    71½"
    Log in for availability
    Log in for availability
    Each
    Each - $452.00
    $452.00
    Each
       $452.00
    Minimum order quantity is 1
    Supplier Number: SWU72K4
  • Upright
    Grey
    Metroseal epoxy
    Steel
    1⅛"
    1⅞"
    84½"
    Log in for availability
    Log in for availability
    Each
    Each - $474.00
    $474.00
    Each
       $474.00
    Minimum order quantity is 1
    Supplier Number: SWU85K4