SmartWall® Single Shelf Support | Avantor
SmartWall® Single Shelf Support
Supplier:   Metropolitan Wire
SmartWall® Single Shelf Support
Supplier:  Metropolitan Wire
Certificates
SmartWall® Single Shelf Support
SmartWall® Single Shelf Support
4 Options Available Below
Product Details & Documents
Create a unique storage and workstation system for often underutilized wall space.
  • Mount directly to slotted uprights
  • Available in lengths to hold 14", 18", 21" and 24" shelves
  • Single shelf supports are used on each end of a single unit or side-by-side unit
  • No tools are necessary to assemble
Suitable for use with Super Erecta®, MetroMax® Q, and MetroMax® i shelves.
Can be used with SmartWall® Uprights to create an easily adjustable wall shelving system.
Metroseal Epoxy finishes feature Microban® antimicrobial product protection which helps to inhibit the growth of microorganisms such as bacteria, mold and fungi which can cause odors, stains and product degradation.
Product Family Options
Informations sur le produit
  • Désignation
    Color
    Finish
    Matière
    Largeur
    Longueur
    Hauteur
    Availability
    Price
  • Single shelf support
    Grey
    Metroseal epoxy
    Steel
    1½"
    16⁹⁄₁₆"
    8³⁄₁₆"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $117.00
    $117.00
    Each
       $117.00
    La quantité minimale de commande est de 1
    Supplier Number: SWS14K4
  • Single shelf support
    Grey
    Metroseal epoxy
    Steel
    1½"
    20⁹⁄₁₆"
    8³⁄₁₆"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $119.00
    $119.00
    Each
       $119.00
    La quantité minimale de commande est de 1
    Supplier Number: SWS18K4
  • Single shelf support
    Grey
    Metroseal epoxy
    Steel
    1½"
    23⁹⁄₁₆"
    8³⁄₁₆"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $124.00
    $124.00
    Each
       $124.00
    La quantité minimale de commande est de 1
    Supplier Number: SWS21K4
  • Single shelf support
    Grey
    Metroseal epoxy
    Steel
    1½"
    26⁹⁄₁₆"
    8³⁄₁₆"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $129.00
    $129.00
    Each
       $129.00
    La quantité minimale de commande est de 1
    Supplier Number: SWS24K4