Tweezers, 3 Wafer Handling, Excelta Corp®
: Excelta
These tweezers are specifically designed handling 3" wafers.
- For scratch free holding of 3" wafers
- Cleanroom safe, anti-magnetic stainless steel
- Various configurations for various applications
The wafer tweezer for 3" wafers are made of stainless steel and are anti-magnetic. They come in a variety of configurations. The 'B' version features a 20° bend in the body to improve handling. The overall length is 4.75".
Frequently Bought Together