SmartWall® Wall Tracks
SmartWall® Wall Tracks
Supplier:   Metropolitan Wire
SmartWall® Wall Tracks
Supplier:  Metropolitan Wire
Certificates
SmartWall® Wall Tracks
SmartWall® Wall Tracks
3 Options Available Below
Product Details & Documents
Create a unique storage and workstation system for underutilized wall space with SmartWall®.
  • Minimum one wall track required per SmartWall® system
  • Joiner plates and stop fastener hardware included
  • Does not include hardware to mount wall tracks to the wall
  • Available in widths of 40, 56 and 72"
Tracks can be used to create a customized wall-mount shelving system by being paired with uprights, grids, and supports. The Metroseal Epoxy finish will prevent the tracks from corrosion and rust, lengthening the life of the product.
Product Family Options
Product Information
  • Désignation
    Color
    Finish
    Matière
    Largeur
    Profondeur
    Hauteur
    Availability
    Price
  • Wall track
    Grey
    Metroseal epoxy
    Steel
    40"
    ¾"
    1⁹⁄₁₆"
    Log in for availability
    Log in for availability
    Each
    Each - $130.00
    $130.00
    Each
       $130.00
    Minimum order quantity is 1
    Supplier Number: SW40K4
  • Wall track
    Grey
    Metroseal epoxy
    Steel
    56"
    ¾"
    1⁹⁄₁₆"
    Log in for availability
    Log in for availability
    Each
    Each - $142.00
    $142.00
    Each
       $142.00
    Minimum order quantity is 1
    Supplier Number: SW56K4
  • Wall track
    Grey
    Metroseal epoxy
    Steel
    72"
    ¾"
    1⁹⁄₁₆"
    Log in for availability
    Log in for availability
    Each
    Each - $160.00
    $160.00
    Each
       $160.00
    Minimum order quantity is 1
    Supplier Number: SW72K4