SmartWall® Wire Grids, Metro™
SmartWall® Wire Grids, Metro™
SmartWall® Wire Grids, Metro™
SmartWall® Wire Grids, Metro™
Metro® SmartWall® Wire Grids create storage and workstation systems for underutilized wall space.
  • Grids may attach directly to SmartWall® uprights
  • No tools are necessary
  • Grids may not overhang the uprights by more than 6" (152 mm)
  • Grid openings measure approximately 3×3" (76×76 mm)
When SmartWall® uprights are not used, select the appropriate grid bracket kit based on the configuration.
Metroseal Gray epoxy finish comes with Microban® antimicrobial product protection which adds a level of protection against bacteria, mold and mildew that can cause stains, odors, and product degradation.
Product Family Options
  • Désignation
    Color
    Finish
    Matière
    Largeur
    Profondeur
    Hauteur
  • Wire grid
    Grey
    Metroseal epoxy
    Steel
    30"
    ½"
    18"
    Each
    Each - $129.00
    $129.00
    Each
       $129.00
     : WG1830K4
  • Wire grid
    Grey
    Metroseal epoxy
    Steel
    36"
    ½"
    18"
    Each
    Each - $137.00
    $137.00
    Each
       $137.00
     : WG1836K4
  • Wire grid
    Grey
    Metroseal epoxy
    Steel
    48"
    ½"
    18"
    Each
    Each - $170.00
    $170.00
    Each
       $170.00
     : WG1848K4
  • Wire grid
    Grey
    Metroseal epoxy
    Steel
    60"
    ½"
    18"
    Each
    Each - $211.00
    $211.00
    Each
       $211.00
     : WG1860K4
  • Wire grid
    Grey
    Metroseal epoxy
    Steel
    30"
    ½"
    24"
    Each
    Each - $222.00
    $222.00
    Each
       $222.00
     : WG2430K4
  • Wire grid
    Grey
    Metroseal epoxy
    Steel
    36"
    ½"
    24"
    Each
    Each - $235.00
    $235.00
    Each
       $235.00
     : WG2436K4
  • Wire grid
    Grey
    Metroseal epoxy
    Steel
    48"
    ½"
    24"
    Each
    Each - $291.00
    $291.00
    Each
       $291.00
     : WG2448K4
  • Wire grid
    Grey
    Metroseal epoxy
    Steel
    36"
    ½"
    30"
    Each
    Each - $237.00
    $237.00
    Each
       $237.00
     : WG3036K4
  • Wire grid
    Grey
    Metroseal epoxy
    Steel
    48"
    ½"
    30"
    Each
    Each - $291.00
    $291.00
    Each
       $291.00
     : WG3048K4