Super Erecta® Wall Mount Shelving End Post Bracket
Wall mount shelving end post bracket, Grey, Metroseal epoxy, Steel, 1¹⁵⁄₁₆", 3⁵⁄₃₂", 1⅞", 250 lbs.
Catalog # 76463-108
Supplier: Metropolitan Wire
Supplier #: BESK4
Jump to:
Product Details & Documents
Replacement end bracket for post-type mount wall shelving.
- Two end brackets are required per post
Corrosion resistant Metroseal Gray epoxy finish features Microban® antimicrobial production protection to keep the product 'cleaner between cleanings'.
Specifications
- Profondeur:3⁵⁄₃₂"
- Finish:Metroseal epoxy
- Load Capacity:250 lbs.
- Largeur:1¹⁵⁄₁₆"
- Color:Grey
- Désignation:Wall mount shelving end post bracket
- Hauteur:1⅞"
- Matière:Steel