Super Erecta® Stackable Ledge for Wire Shelving, 4" High
Super Erecta® Stackable Ledge for Wire Shelving, 4" High
Supplier:   Metropolitan Wire
Super Erecta® Stackable Ledge for Wire Shelving, 4" High
Supplier:  Metropolitan Wire
Certificates
Super Erecta® Stackable Ledge for Wire Shelving, 4" High
Super Erecta® Stackable Ledge for Wire Shelving, 4" High
11 Options Available Below
Product Details & Documents
Prevent items from protruding or falling from shelves.
  • Only compatible with wire shelving
  • Ledges may be stacked to form an 8" (203 mm) or higher barrier
Corrosion resistant Metroseal Gray epoxy finish features Microban® antimicrobial production protection to keep the product 'cleaner between cleanings'.
Product Family Options
Product Information
  • Désignation
    Color
    For Use With
    Finish
    Matière
    Largeur
    Longueur
    Hauteur
    Availability
    Price
  • Metroseal gray epoxy ledge
    Grey
    Super Erecta shelving
    Metroseal epoxy
    Steel
    1⅝"
    14"
    4"
    Log in for availability
    Log in for availability
    Each
    Each - $51.80
    $51.80
    Each
       $51.80
    Minimum order quantity is 1
    Supplier Number: L14N-4K4
  • Metroseal gray epoxy ledge
    Grey
    Super Erecta shelving
    Metroseal epoxy
    Steel
    1⅝"
    18"
    4"
    Log in for availability
    Log in for availability
    Each
    Each - $53.90
    $53.90
    Each
       $53.90
    Minimum order quantity is 1
    Supplier Number: L18N-4K4
  • Metroseal gray epoxy ledge
    Grey
    Super Erecta shelving
    Metroseal epoxy
    Steel
    1⅝"
    21"
    4"
    Log in for availability
    Log in for availability
    Each
    Each - $56.70
    $56.70
    Each
       $56.70
    Minimum order quantity is 1
    Supplier Number: L21N-4K4
  • Metroseal gray epoxy ledge
    Grey
    Super Erecta shelving
    Metroseal epoxy
    Steel
    1⅝"
    24"
    4"
    Log in for availability
    Log in for availability
    Each
    Each - $61.60
    $61.60
    Each
       $61.60
    Minimum order quantity is 1
    Supplier Number: L24N-4K4
  • Metroseal gray epoxy ledge
    Grey
    Super Erecta shelving
    Metroseal epoxy
    Steel
    1⅝"
    30"
    4"
    Log in for availability
    Log in for availability
    Each
    Each - $69.30
    $69.30
    Each
       $69.30
    Minimum order quantity is 1
    Supplier Number: L30N-4K4
  • Metroseal gray epoxy ledge
    Grey
    Super Erecta shelving
    Metroseal epoxy
    Steel
    1⅝"
    36"
    4"
    Log in for availability
    Log in for availability
    Each
    Each - $77.70
    $77.70
    Each
       $77.70
    Minimum order quantity is 1
    Supplier Number: L36N-4K4
  • Metroseal gray epoxy ledge
    Grey
    Super Erecta shelving
    Metroseal epoxy
    Steel
    1⅝"
    42"
    4"
    Log in for availability
    Log in for availability
    Each
    Each - $94.50
    $94.50
    Each
       $94.50
    Minimum order quantity is 1
    Supplier Number: L42N-4K4
  • Metroseal gray epoxy ledge
    Grey
    Super Erecta shelving
    Metroseal epoxy
    Steel
    1⅝"
    48"
    4"
    Log in for availability
    Log in for availability
    Each
    Each - $94.50
    $94.50
    Each
       $94.50
    Minimum order quantity is 1
    Supplier Number: L48N-4K4
  • Metroseal gray epoxy ledge
    Grey
    Super Erecta shelving
    Metroseal epoxy
    Steel
    1⅝"
    54"
    4"
    Log in for availability
    Log in for availability
    Each
    Each - $102.00
    $102.00
    Each
       $102.00
    Minimum order quantity is 1
    Supplier Number: L54N-4K4
  • Metroseal gray epoxy ledge
    Grey
    Super Erecta shelving
    Metroseal epoxy
    Steel
    1⅝"
    60"
    4"
    Log in for availability
    Log in for availability
    Each
    Each - $102.00
    $102.00
    Each
       $102.00
    Minimum order quantity is 1
    Supplier Number: L60N-4K4
  • Metroseal gray epoxy ledge
    Grey
    Super Erecta shelving
    Metroseal epoxy
    Steel
    1⅝"
    72"
    4"
    Log in for availability
    Log in for availability
    Each
    Each - $122.00
    $122.00
    Each
       $122.00
    Minimum order quantity is 1
    Supplier Number: L72N-4K4