Orbit DNA Model | Avantor
Orbit DNA Model
Single model
Catalog # 470325-252
Supplier:   Cochranes of Oxford
Fournisseur #:  0374
Orbit DNA Model
Catalog # 470325-252
Orbit DNA Model
Orbit DNA Model
Single model
Price
$18.65
Log in pour la disponibilité
Kit of 1
Product Details & Documents
Understand DNA/RNA structure.
  • Available as a single kit, or a class pack of 12
  • Small kits can be combined to form larger sequences
This Orbit molecular model aids understanding of the process of DNA/RNA transcription.
The model shows six base pairs with each base (guanine, cytosine, thymine, adenine, and uracil) in a different color and the sugar rings, phosphate groups, and hydrogen bonds in black, red, and white respectively. The Class pack can also be combined for a series of 72 base pairs.
The strands of DNA can be joined and taken apart to demonstrate the transcription of RNA.
Spécifications
  • Désignation:
    Single model
  • Cat. No.:
    470325-252