Hughes Portable, Self-Contained, 10 gal. Gravity-Fed Eyewash Station, Justrite®
Fournisseur: Justrite
Certificats
À propos de cet article
Portable design with carrying handles and concealed wall mounting bracket make moving and relocating easy.
- Wall mounted
- High-visibility orange color is easy to identify in emergency
- Simple to use with pull-down activation handle
- Wide fill opening enables quick inspection, cleaning, and filling
The Hughes portable, self-contained 10 gal. gravity-fed eyewash station is suitable wherever continuous water isn't available, with no plumbing needed. The gravity-fed eyewash supplies a continuous, controlled flow that exceeds the ANSI Z358.1 standard of 1.5 LPM (0.4 GPM) for 15 minutes, providing a reliable safety solution no matter where it is located. Concealed mounting bracket make moving and relocating easy. High-visibility color is easy to identify in emergency situations. Simple to use with pull-down activation handle.
Mounting type: Wall bracket
Operating time: 15 minutes
Certifications: UL classified to ANSI Z358.1-2014. ANSI Z358.1-2014 and EN 15154 compliant.
Spécifications
- Catalog No:
- 76462-628
- 76462-630
- Ambient operating temperature:
- 5...35 °C (41...95 °F)
- 5...35 °C (41...95 °F)