SP Wilmad-LabGlass Stem Coaxial Small Volume NMR Inserts, SP Industries
Fournisseur: SP Industries
Certificats
À propos de cet article
The most versatile and reliable coaxial system available for NMR experiments.
- Manufactured from ASTM Type 1 class A glass, ideal for variable temperature studies
- Outer tube must be ordered separately depending on magnetic field strength
General applications include small volume NMR, external referencing, external locking and magnetic susceptibility determination.
Frequently Bought Together