SP Wilmad-LabGlass High Temperature EPR Dewars for Bruker® ER4114HT Cavity, SP Industries
Fournisseur: SP Industries
Certificats
À propos de cet article
This X band EPR sample tube was made specifically for Bruker’s ER 4114HT-1000 cavity.
- Provides low temperature gradients in the sample region
- Maximum working temperature is 1200 K
- Use with Wilmad's 750-PQ-7.1 sample tubes
- Made of Suprasil
The three wall finger system made from synthetic quartz yields low thermal expansion coefficient.
Frequently Bought Together