Spigot, LDPE
Spigot, LDPE, 3.3×1.15×2.8", 3/4"
470330-666
: Dynalon
Supplier #: 612935
Standard drum spigot.
Plastic spigots are molded of low-density polyethylene of white with 3/4" IPS thread. Plastic spigots have no gaskets or inside threads to leak. Will fit standard threaded drums.
- Désignation:Spigot
- Matière:LDPE
- L×W×H:3.3×1.15×2.8"
- Epaisseur:3/4"