Face Shield
Product Details & Documents
Face shield provides reliable splash protection in a variety of applications and environments.
- Covers the entire face
- Non sterile
- Anti-fog coating
- Manufactured in an FDA registered facility
Safe, comfortable, and easy to use. This facemask can be worn with or without goggles or glasses. It offers reliable protection and features a soft elastic band, padded headband, and clear PET shield. Each shield is individually wrapped, and can either be disposed of or sanitized.
Spécifications
- Désignation:Face shield
- Matière:PET