Slim T™ Buckets, Micronova
Supplier: Micronova
Certificates
About this item
Rectangular, 6 gallon buckets with a 19" interior width are an ideal choice for use with a variety of flat head mops and string mops in controlled environments. Available with or without heavy-duty 3" casters.
- 6 Gallon capacity
- Volume graduations with gallon and liter marks
- Color-coded with a choice of red, yellow, blue, green, and grey
- B-7: Use as replacement buckets for SlimLine™ and Slim T™ series carts
- B-7AC: Use as replacements in BucketBinder™ multi bucket systems
- Autoclavable
Micronova Slim T™ polypropylene buckets are designed for stand-alone use or as replacements for SlimLine™ or Slim T™ series carts. They also work well with BucketLiners™ and the range of W-9 series wringers. Buckets with casters, use as a single bucket system, or as replacements in BucketBinder™ multi bucket systems.
Frequently Bought Together