Silicon Nitride TEM Window Grids, Electron Microscopy Sciences
À propos de cet article
Silicon Nitride TEM Window Grids perform well under harsh lab conditions.
- Plasma cleanable - can be vigorously plasma cleaned to remove organic contamination
- Field to field uniformity - less than 0.5 nm variation in film thickness across an entire production log, not just a single window grid
- Tolerates temperatures above 1000 °C - supports use in environmental TEMs where dynamic processes are observed at high temperatures
- Withstands harsh conditions - provides an ideal balance of imaging resolution, chemical stability and mechanical strength
- Incorporates LPCVD, low-stress(~250 MPa), non-stoichiometric silicon nitride to provide flat, insulating and hydrophobic surfaces
The silicon frames are 100 µm thick. Grids fit standard 3 mm holders and most double tilt holders. They come in clear gel-boxes for simpler sample preparation.
3 Options Available Below
Product Details & Documents
Silicon Nitride TEM Window Grids perform well under harsh lab conditions.
- Plasma cleanable - can be vigorously plasma cleaned to remove organic contamination
- Field to field uniformity - less than 0.5 nm variation in film thickness across an entire production log, not just a single window grid
- Tolerates temperatures above 1000 °C - supports use in environmental TEMs where dynamic processes are observed at high temperatures
- Withstands harsh conditions - provides an ideal balance of imaging resolution, chemical stability and mechanical strength
- Incorporates LPCVD, low-stress(~250 MPa), non-stoichiometric silicon nitride to provide flat, insulating and hydrophobic surfaces
The silicon frames are 100 µm thick. Grids fit standard 3 mm holders and most double tilt holders. They come in clear gel-boxes for simpler sample preparation.
Product Family Options
Silicon Nitride TEM Window
Informations sur le produit
- UtilisationDimensionsÉpaisseurAvailabilityPrice
- High resolution imaging25 µm5 nmDiscontinued-Supplier Number: 76042-43Spécifications:MarketsourceMarketsource
- High resolution imaging(8) 50 µm, (1) 50×100 µm5 nmDiscontinued-Supplier Number: 76042-44Spécifications:MarketsourceMarketsource
- High resolution imaging(2) 50×1500 µm5 nmDiscontinued-Supplier Number: 76042-45Spécifications:MarketsourceMarketsource
- Robust, increased high resolution(8) 100 µm, (1) 100×350 µm10 nmLog in pour la disponibilitéPack of 10$838.20Supplier Number: 76042-46
- —(8) 250 µm, (1) 250×500 µm10 nmDiscontinued-Supplier Number: 76042-47Spécifications:MarketsourceMarketsource
- Supplier Number: 76042-48
- Everyday500 µm20 nmDiscontinued-Supplier Number: 76042-49Spécifications:MarketsourceMarketsource
- Everyday and demanding conditions(9) 100 µm50 nmDiscontinued-Supplier Number: 76042-50Spécifications:MarketsourceMarketsource
- Demanding conditions100 µm50 nmDiscontinued-Supplier Number: 76042-53Spécifications:MarketsourceMarketsource
- Supplier Number: 76042-52
- Demanding conditions1000 µm50 nmDiscontinued-Supplier Number: 76042-51Spécifications:MarketsourceMarketsource