Graphene Oxide TEM Support Films, Electron Microscopy Sciences
À propos de cet article
Graphene Oxide (GO) support film is a super thin (<1 nm), naturally hydrophilic layer placed over the Holey, Lacey or Quantifoil support film on copper or gold grids.
- Works well with Holey Carbon, Lacey Carbon and Quantifoil grid types, effectively spanning the gaps
- Less expensive to produce due to complexity of graphene manufacturing
- Better background contrast than graphene, results in higher resolution
Pre-treatment of GO Support Films is unnecessary. By default, the hydrophilic surface spreads particles evenly across the grid. A hydrophobic surface can be achieved by heating in the air. Note: plasma cleaning or glow discharge will damage the support film.
11 Options Available Below
Product Details & Documents
Graphene Oxide (GO) support film is a super thin (<1 nm), naturally hydrophilic layer placed over the Holey, Lacey or Quantifoil support film on copper or gold grids.
- Works well with Holey Carbon, Lacey Carbon and Quantifoil grid types, effectively spanning the gaps
- Less expensive to produce due to complexity of graphene manufacturing
- Better background contrast than graphene, results in higher resolution
Pre-treatment of GO Support Films is unnecessary. By default, the hydrophilic surface spreads particles evenly across the grid. A hydrophobic surface can be achieved by heating in the air. Note: plasma cleaning or glow discharge will damage the support film.
Product Family Options
Graphene Oxide on Holey Carbon
Informations sur le produit
- MeshGrid TypeAvailabilityPrice
- Supplier Number: GOHC300CU10
- 300CopperDiscontinued-Supplier Number: GOHC300CU25Spécifications:MarketsourceMarketsource
- 300CopperDiscontinued-Supplier Number: GOHC300CU50Spécifications:MarketsourceMarketsource
Graphene Oxide on Lacey Carbon
Informations sur le produit
- MeshGrid TypeAvailabilityPrice
- Supplier Number: GOLC300CU10
- 300CopperDiscontinued-Supplier Number: GOLC300CU25Spécifications:MarketsourceMarketsource
- Supplier Number: GOLC300CU50
- 300CopperDiscontinued-Supplier Number: GOLC300CU100Spécifications:MarketsourceMarketsource
Graphene Oxide on Quantifoils R1.2/1.3
Informations sur le produit
- MeshGrid TypeAvailabilityPrice
- Supplier Number: GOQ400R1213A10
- Supplier Number: GOQ400R1213A25
- 400GoldDiscontinued-Supplier Number: GOQ400R1213AU50Spécifications:MarketsourceMarketsource
- Supplier Number: GOQ400R1213C10
- Supplier Number: GOQ400R1213C25
- Supplier Number: GOQ400R1213C50
- 400CopperDiscontinued-Supplier Number: GOQ400R1213100Spécifications:MarketsourceMarketsource
Graphene Oxide on Quantifoils R2/2
Informations sur le produit
Graphene Oxide on Quantifoils R2/4
Informations sur le produit
- MeshGrid TypeAvailabilityPrice
- Supplier Number: GOQ200R24CU10
- 200CopperDiscontinued-Supplier Number: GOQ200R24CU25Spécifications:MarketsourceMarketsource
- 200CopperDiscontinued-Supplier Number: GOQ200R24CU50Spécifications:MarketsourceMarketsource
- 300CopperDiscontinued-Supplier Number: GOQ300R24CU10Spécifications:MarketsourceMarketsource
- 300CopperDiscontinued-Supplier Number: GOQ300R24CU25Spécifications:MarketsourceMarketsource
- 300CopperDiscontinued-Supplier Number: GOQ300R24CU50Spécifications:MarketsourceMarketsource