mPrep/f™ Pipette Filter Couplers, Electron Microscopy Sciences | Avantor
mPrep/f™ Pipette Filter Couplers, Electron Microscopy Sciences
mPrep/f™ Pipette Filter Couplers, Electron Microscopy Sciences
Certificates
mPrep/f™ Pipette Filter Couplers, Electron Microscopy Sciences
mPrep/f™ Pipette Filter Couplers, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Filter couplers prevent the introduction of damaging reagents into pipettors. They also improve the fit of mPrep/g™ capsules on some pipettors.
Available in two pore sizes. Standard mPrep/f30™ has a nominal 30 µm pore size filter appropriate for most applications. Extreme mPrep/f13™ has a nominal 13 µm pore size filter for use with biohazards and very aggressive reagents.
Delivery information: Pack includes 16 filter couplers and a capsule storage box.
Product Family Options
Informations sur le produit
  • Tip Style
    Availability
    Price
  • Filter-couplers, f30 - Standard
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 16
    Pack of 16 - $184.39
    $184.39
    Pack of 16
       $184.39
    Supplier Number: 85010-11
  • Filter-couplers, f13 - Extreme
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 16
    Pack of 16 - $184.39
    $184.39
    Pack of 16
       $184.39
    Supplier Number: 85010-12