Avantor® ACE® UltraCore C18, Analytical HPLC/UHPLC Columns
: Hichrom
Avantor® ACE® UltraCore C18 are high performing solid-core (core-shell) particle columns designed for a wide range of LC applications. This densely bonded octadecyl phase can be used to separate acidic, basic and neutral molecules, based on hydrophobic retention mechanisms.
- High purity, base deactivated silica with excellent reproducibility
- High efficiency separations comparable with sub-2 µm packings but lower back pressure
- Ideal for a wide range of analytes differing in hydrophobicity
These stainless steel columns are available in a variety of dimensions and are highly compatible with LC-MS and high sensitivity applications.
- Catalog No:
- 76458-004
- 76458-006
- 76458-000
- 76458-002
- 76458-012
- 76458-014
- 76458-008
- 76458-010
- 76457-996
- 76457-998
- 76457-992
- 76457-994
- Carbon load:
- 8.2%
- 8.2%
- 8,2%
- 8,2%
- 8.2%
- 8.2%
- 8.2%
- 8.2%
- 8.2%
- 8,2%
- 8.2%
- 8.2%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Max. pressure:
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- pH range:
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- Surface area:
- 115 m²/g
- 115 m²/g
- 115 m²/g
- 115 m²/g
- 115 m²/g
- 115 m²/g
- 115 m²/g
- 115 m²/g
- 115 m²/g
- 115 m²/g
- 115 m²/g
- 115 m²/g
- Temperature limit:
- 60 °C (pH 2); 40 °C (pH 9)
- 60 °C (pH 2); 40 °C (pH 9)
- 60 °C (pH 2); 40 °C (pH 9)
- 60 °C (pH 2); 40 °C (pH 9)
- 60 °C (pH 2); 40 °C (pH 9)
- 60 °C (pH 2); 40 °C (pH 9)
- 60 °C (pH 2); 40 °C (pH 9)
- 60 °C (pH 2); 40 °C (pH 9)
- 60 °C (pH 2); 40 °C (pH 9)
- 60 °C (pH 2); 40 °C (pH 9)
- 60 °C (pH 2); 40 °C (pH 9)
- 60 °C (pH 2); 40 °C (pH 9)
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
Frequently Bought Together