ContactUsLinkComponent
Eisco Economy Microscope Slide Viewer
Catalogue # 470324-796
Fournisseur:  Wards
undefined
Eisco Economy Microscope Slide Viewer
Catalogue # 470324-796
Fournisseur:  Wards
Supplier Number:  BI0276BV
Spécifications
  • Description:
    Economy microscope slide viewer
  • W×D×H:
    6.6×3.9×7.5"
  • Cat. no.:
    470324-796
Spécifications
À propos de cet article
A fun introduction to the world of microscopy for students.
  • Lightweight and easy to use
  • Uses ambient light
  • Wireless, no battereies or light bulbs required
  • Compact design for easy storage and handling
This economical microscope slide viewer is light weight and economical. The unit does not require batteries or to be plugged into to an electrical source. Light from a lamp or natural light can be directed to the white surface and the light will reflect into the viewer.