dGTP Solution
dGTP solution 100 mM, 25 µmol
Catalog # CA76459-520
New Product
Supplier: New England Biolabs (NEB)
Supplier #: N0442S
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
dGTP Solution contains 0.25 ml of 100 mM ultrapure dGTP as a sodium salt at pH 7.5.
- ≥99% purity as determined by HPLC analysis
- Used in ligation independent cloning
- Also available in solution mix or set format
It is supplied in Ultrapure water as a sodium salt at pH 7.5. The dGTP Solution is supplied at a concentration of 100 mM (±5 mM) as determined by A260 absorbance. Can be diluted using sterile distilled water, preferably Milli-Q® water or can be diluted using sterile TE (10 mM Tris-HCl, 1 mM EDTA (pH 7.5)).
Specifications
Specifications
- Désignation:dGTP solution 100 mM
- Taille:25 µmol