À propos de cet article
Improve the microbial barrier and cleanliness of your sterilization wrap.
- Manufactured and packaged in a controlled cleanroom environment
- Autoclavable at 121 to 125 °C
- Tear and puncture resistant
- Hydrophobic
- Lint-free
Round shaped Tyvek® Wrappers are an ideal way to cover round openings for steam sterilization. Eliminate the wasted time and expense of cutting larger square sheets of sterilization wrap to desired shapes and sizes. Clean cut circles delivered in poly bags ensure cleanliness. Simply secure in place with autoclaveable elastic bands.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.