DuPont™ Tyvek® Mini-Muff, Keystone Cleanroom Products
Save time and standardize the activities in equipment preparation for sterilization.
- Manufactured and packaged in a controlled cleanroom environment
- Reduces labor costs
- Simplifies the conformance to wrapping SOP's
- Autoclavable at 121 to 125 °C
- Tear and puncture resistant
- Hydrophobic
- Lint-free
- Conveniently packaged-100 per sealed poly bag
Save time and standardize the activities in equipment preparation for sterilization. Tyvek® Muffs provide a convenient means to cover flasks, beakers, containers, media bottles, valves, pipes and hoses. These closures are breathable and can be used to prevent particulates and microbial contamination from entering openings of any size. Since they are autoclavable, they can be used during sterilization to reduce time consuming wrapping of open containers. Each closure incorporates latex-free rubber elastic to create a positive seal around round shaped openings.
: Certificates of conformance sent with each shipment.
Frequently Bought Together