Isolator Bags, Self-Sealing Tyvek® 1073B/ PE Steam Sterilization Chevron Peel Pouch with Steam Indicator and Hang Hole, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
Certificates
About this item
Improve the prep time, performance, microbial barrier, and cleanliness of your autoclave peel pouches.
- Manufactured and packaged in a controlled cleanroom environment
- Integrated Hang Hole is ideal for use with Isolator VHP decontamination cycles
- Easy, fast, and reliable self-sealing closure system
- Autoclavable at 121 to 125 °C
- Peels open cleanly and lint-free
- Chevron peel and hydrophobic
- Tear and puncture resistant
- Integrated Steam Sterilization Indicator
A significant upgrade over paper-based steam sterilization pouches. Integrated Hang-Hole for use in Isolators. These Tyvek® peel pouches will not rip or puncture like paper-based autoclave pouches routinely do. Save valuable time in equipment preparation with the easy-to-use self-sealing closure system.
No need for the use and validation of a heat sealer. The medical Grade 1073B Tyvek® material has the highest microbial barrier properties of any steam sterilization compatible materials. These pouches are also compatible with VHP Decontamination cycles commonly used with Isolator technology. Will not absorb or adsorb H₂O₂.
Frequently Bought Together