Tyvek® 1073B PE Steam Sterilization Tubing, Continuous Form, Keystone Cleanroom Products
Improve the prep time, performance, microbial barrier and cleanliness of your autoclave peel pouches.
- Manufactured and packaged in a controlled cleanroom environment
- Sizing flexibility and hydrophobic
- Plastic core (no corrugated material)
- Unroll and cut to desired length, heat seal both ends
- Autoclavable at 121 to 125 °C
- Peels open cleanly, lint-free
- Tear and puncture resistant
A significant upgrade over paper based steam sterilization tubing . These Tyvek® peel pouches will not rip or puncture like paper based autoclave pouches routinely do. The medical Grade 1073B Tyvek® material has the highest microbial barrier properties of any steam sterization compatible materials. These pouches are also compatible with VHP Decontamination cycles commonly used with Isolator technology. Will not absorb or adsorp H₂O₂.
Frequently Bought Together