Tyvek® Stopper Bags, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
Certificates
About this item
1073B Tyvek® / HDPE autoclave bags are ideal for steam sterilization of pharmaceutical components such as vial and syringe stoppers and caps. Also a good choice for heavy equipment parts where visibility into the bag is required.
- Manufactured and packaged in a controlled cleanroom environment
- Steam sterilization indicator - autoclavable at 121 to 125 °C
- Tear and puncture resistant
- Hydrophobic and lint-free
- Semi-transparent HDPE film side
- Excellent impact resistance with exceptional microbial barrier
- Consistently strong, double welded factory seals
1073B Tyvek® side permits steam penetration during sterilization. These are a good choice for heavy, or pharmaceutical change parts with sharp corners or where visibility into the bag is required after sterilization.
Delivery: Supplied with Certificates of Conformance.
Frequently Bought Together