BHD Autoclave Bags with Self-Seal Closure, Blue, Keystone Cleanroom Products
Self-sealing BHD Autoclave bags can be used as either the inner, or outer bag for the steam sterilization of pharmaceutical change parts.
- Manufactured and packaged in a controlled cleanroom environment
- Reduces labor costs
- Simplifies the conformance to wrapping SOP's
- Autoclavable up to 135 °C
- Tear and puncture resistant
- Significant process improvement compared to wrapping with BioShield paper
- Eliminates need to cut material and the use of autoclave tape
Standardize and simplify the way equipment is wrapped for steam sterilization.
The use of these 2 ply self-sealing bags will also save valuable time in the equipment prep area. The 2-ply BHD material resists ripping and punctures.
Since 100% of the surface area allows for steam penetration and drying, each load can be configured for maximum efficieny and throughput.
Frequently Bought Together