About this item
BHD Autoclave bags with Tyvek® Drawstrings can be used as either the inner, or outer bag for the steam sterilization of pharmaceutical change parts.
- Manufactured and packaged in a controlled cleanroom environment
- Reduces labor costs
- Simplifies the conformance to wrapping SOP's
- Steam sterilization indicator - autoclavable up to 135 °C
- Tear and puncture resistant
- Significant process improvement compared to wrapping with BioShield paper
- Eliminates need to cut material and the use of autoclave tape
Standardize and simplify the way equipment is wrapped for steam sterilization.
The use of these 2 ply drawstring bags will also save valuable time in the equipment prep area. The 2-ply BHD material resists ripping and punctures.
Since 100% of the surface area allows for steam penetration and drying, each load can be configured for maximum efficieny and throughput.
The drawstring bags can be aseptically opened without generating particles like cutting other types of bags open can.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.