Tyvek® 1422A Steam Sterilization Tubing, Continuous Form, Keystone Cleanroom Products
Fournisseur: KEYSTONE ADJUSTABLE CAP CO., INC.
Certificats
À propos de cet article
Continuous Form tubing made from 1422A Tyvek® provides the user with tremendous sizing flexibility when wrapping pharmaceutical equipment for steam sterilization.
- Manufactured and packaged in a controlled cleanroom environment
- On a plastic core (no corrugated material)
- Tear and puncture resistant
- Hydrophobic
- Lint-free
- Consistently strong factory seals
- Exceptional microbial barrier
This steam sterilization tubing is a great choice when there is a wide range of equipment parts that need to be wrapped for sterilization.
Simply unroll the tubing to the desired length, slide the equipment into the tube and then heat seal each end closed.
The drapeable nature of the 1422A Tyvek® material is ideal for wrapping equipment parts where one may need to 'feel through' the bag; especially when lifting, or handling the wrapped part.
Since 100% of the surface area allows for steam penetration and drying, each load can be configured for maximum efficiency and throughput.
Delivery: Certificates of Conformance sent with each shipment.
Frequently Bought Together