Avantor® ACE® UltraCore C18-Amide, Analytical HPLC/UHPLC Columns
Fournisseur: Hichrom
Certificats
À propos de cet article
Avantor® ACE® UltraCore C18-Amide are high performing solid-core (core shell) particle columns designed for a range of reversed-phase chromatographic applications. Additional mechanisms of interaction such as H-bonding give an alternative selectivity for moderately polar analytes.
- High purity, base deactivated silica with excellent reproducibility
- Can retain more polar compounds in 100% aqueous mobile phase
- Alternative selectivity to alkyl phases
These stainless steel columns are available in a variety of dimensions and are highly compatible with LC-MS and high sensitivity applications.
Spécifications
- Catalog No:
- 76458-042
- 76458-044
- 76458-060
- 76458-040
- 76458-062
- 76458-048
- 76458-050
- 76458-046
- 76458-058
- 76458-052
- 76458-054
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Max. pressure:
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- pH range:
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- Surface area:
- 115 m²/g
- 115 m²/g
- 115 m²/g
- 115 m²/g
- 115 m²/g
- 115 m²/g
- 115 m²/g
- 115 m²/g
- 115 m²/g
- 115 m²/g
- 115 m²/g
- Temperature limit:
- 60 °C (pH 2); 40 °C (pH 9)
- 60 °C (pH 2); 40 °C (pH 9)
- 60 °C (pH 2); 40 °C (pH 9)
- 60 °C (pH 2); 40 °C (pH 9)
- 60 °C (pH 2); 40 °C (pH 9)
- 60 °C (pH 2); 40 °C (pH 9)
- 60 °C (pH 2); 40 °C (pH 9)
- 60 °C (pH 2); 40 °C (pH 9)
- 60 °C (pH 2); 40 °C (pH 9)
- 60 °C (pH 2); 40 °C (pH 9)
- 60 °C (pH 2); 40 °C (pH 9)
- USP code:
- L60
- L60
- L60
- L60
- L60
- L60
- L60
- L60
- L60
- L60
- L60