Avantor® ACE® UltraCore Biphenyl, Analytical HPLC/UHPLC Columns
Supplier: Hichrom
Certificates
About this item
Avantor® ACE® UltraCore Biphenyl are high performing solid-core (core-shell) particle columns, designed for a range of chromatographic applications and is especially well suited for retention of aromatic compounds that elute early on alkyl phases.
- High purity, base deactivated silica with excellent reproducibility
- Ideal for analytes with π-bonding, electron withdrawing or delocalised groups
- Alternative selectivity to alkyl phases and traditional phenyl phases
These stainless steel columns are available in a variety of dimensions and are highly compatible with LC-MS and high sensitivity applications.
Specifications
- Catalog No:
- 76458-020
- 76458-022
- 76458-016
- 76458-038
- 76458-018
- 76458-024
- 76458-026
- 76458-028
- 76458-034
- 76458-036
- 76458-030
- 76458-032
- Carbon load:
- 6.5%
- 6.5%
- 6.5%
- 6.5%
- 6.5%
- 6.5%
- 6.5%
- 6.5%
- 6.5%
- 6.5%
- 6.5%
- 6.5%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Max. pressure:
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.56 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.56 psi)
- 600 bar (8702.56 psi)
- pH range:
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- Surface area:
- 115 m²/g
- 115 m²/g
- 115 m²/g
- 115 m²/g
- 115 m²/g
- 115 m²/g
- 115 m²/g
- 115 m²/g
- 115 m²/g
- 115 m²/g
- 115 m²/g
- 115 m²/g
- Temperature limit:
- 60 °C (pH 2); 40 °C (pH 9)
- 60 °C (pH 2); 40 °C (pH 9)
- 60 °C (pH 2); 40 °C (pH 9)
- 60 °C (pH 2); 40 °C (pH 9)
- 60 °C (pH 2); 40 °C (pH 9)
- 60 °C (pH 2); 40 °C (pH 9)
- 60 °C (pH 2); 40 °C (pH 9)
- 60 °C (pH 2); 40 °C (pH 9)
- 60 °C (pH 2); 40 °C (pH 9)
- 60 °C (pH 2); 40 °C (pH 9)
- 60 °C (pH 2); 40 °C (pH 9)
- 60 °C (pH 2); 40 °C (pH 9)
- USP code:
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11