ContactUsLinkComponent
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    EDX-XEDS TEM window grid
  • Environmentally Preferable:
  • Cat. no.:
    76439-544
Suspended germanium provides a unique calibration standard for x-ray energy dispersive spectroscopy. Since Ge is not typically found in TEM columns, the calibration samples provide a material that cannot be mistaken for instrument components and their signal peaks. The regime in which system peaks normally occur (2 to 9 keV and 11 to 20 keV) is devoid of peaks from the Ge.
  • 20 nm thick germanium (Ge) coating on microporous 20 nm thick, low-stress silicon nitride (SiN)
  • 2 µm pores on 1:1 pitch grid pattern
  • 100 µm thick frame, fits 3 mm sample holders
  • (1) 500×500 µm window
The Ge is suspended across 2 µm pores that are patterned on a grid of 20 nm thick silicon nitride.
The single 500×500 µm window is compatible with high tilt angle tomography, since at 70° of tilt, the thin and beveled 100 µm silicon frame allows you to use a ~50×50 µm region within the center of the window from any rotational orientation.
These EDX calibration standards were developed in partnership with Dr. Nestor J. Zaluzec from the Electron Microscopy Center and the Center for Nanoscale Materials at Argonne National Laboratory.
Applications in detector energy axis and energy resolution calibration, detector window transmission evaluation, detector solid angle measurements, electron optical instrument system peak measurements, and specimen.