X-Ray Windows are the ideal substrates for x-ray microscopy and x-ray spectroscopy techniques. State-of-the-art manufacturing and expert engineering allow for competitive prices of these windows. Made in the USA, these X-Ray windows are entirely plasma cleanable to remove organic contamination. Flat, uniformly deposited films provide consistent backgrounds with low field-to-field variability and high x-ray transmission.
- Available with silicon nitride or silicon oxide membranes
Low-stress LPCVD Silicon nitride membranes are mechanically strong and well-suited for high temperature and differential pressure environments. The frame is 320 µm thick and non-porous.
Proprietary wrinkle-free G-FLAT™ Silicon oxide membranes are well-suited for correlative optical and x-ray microscopy and analyses requiring a nitrogen-free background. suited for biological imaging studies, with a glass-like hydrophilic surface. They have a 310 µm thick frame and unique G-Flat™ wrinkle-free silicon oxide film. They are compatible with ultra-high vacuum (UHV) applications (300 nm membrane) and non-porous.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.