Snap It Ampoule Opener, Electron Microscopy Sciences
Snap It Ampoule Opener, Electron Microscopy Sciences
Snap It Ampoule Opener, Electron Microscopy Sciences
Certificates
Snap It Ampoule Opener, Electron Microscopy Sciences
Snap It Ampoule Opener, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Available in regular (1 to 15 ml) and large (5 to 25 ml) sizes.
Product Family Options
Product Information
  • Désignation
    For
    Availability
    Price
  • Regular Size
    1 - 15 ml ampoules
    Log in for availability
    Log in for availability
    Box of 10
    Box of 10 - $561.39
    $561.39
    Box of 10
       $561.39
    Supplier Number: 72054-80
  • Large Size
    5 - 25 ml ampoules
    Log in for availability
    Log in for availability
    Box of 10
    Box of 10 - $582.73
    $582.73
    Box of 10
       $582.73
    Supplier Number: 72054-81