EMS Grid Prep Holders, Electron Microscopy Sciences | Avantor
EMS Grid Prep Holders, Electron Microscopy Sciences
Marketsource
EMS Grid Prep Holders, Electron Microscopy Sciences
Marketsource
Certificates
EMS Grid Prep Holders, Electron Microscopy Sciences
EMS Grid Prep Holders, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
EMS preparation tray may be use for staining and/or placing an evaporator for the coating of grids.
  • Medical grade aluminum
  • Includes a plastic storage box
These unique trays have a M4 tapped hole in the center which allows for insertion of pin mount. They also have a screw pin in the middle that may be used as a handle for manipulating the trays.
Product Family Options
Informations sur le produit
  • Désignation
    Dimensions
    Availability
    Price
  • Grid prep holder, capacity 25, thickness 6.4 mm
    35×35 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $456.11
    $456.11
    Each
       $456.11
    Supplier Number: 71175-00
  • Grid prep holder, capacity 36, thickness 6 mm
    43×43 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $304.38
    $304.38
    Each
       $304.38
    Supplier Number: 71175-01
  • Grid prep holder, capacity 81, thickness 6 mm
    65×65 mm
    Discontinued
    Supplier Number: 71175-02
    Spécifications:
    Marketsource
    Marketsource