Eisco Glass Weighing Bottles with Stopper, Tall Form
Supplier: Wards
Certificates
About this item
Durable, high quality glass weighing bottle for a variety of laboratory uses.
- Borosilicate 3.3 glass
- Tall form
- Autoclavable
- Superior chemical durability
- Low temperature gradient- can withstand high temperatures and thermal shocks
Eisco Labs Borosilicate glass has the following heat tolerances: strain point –515 °C; annealing point 565 °C and softening point 820 °C.
Delivery: Includes interchangable ground glass stopper.
Frequently Bought Together