Luna™ 2.0 Interactive Projection Camera, Learning Resources SE
Interactive Projection Camera, 3 MP
Catalog # 470330-652
Supplier: Learning Resources
: LER4427
Luna™ 2.0 Interactive Projection Camera, Learning Resources SE
Catalog # 470330-652
Supplier: Learning Resources
Jump to:
Product Details & Documents
Affordable document camera for virtual learning.
- 3 MP digital images
- Adjustable gooseneck
- Autofocus eight LED lights
- Internal microphone
Contemporary new design and enhanced image quality meets reliable simplicity. Easy-to-use digital projection camera is also a webcam and photo/video camera. Great for students, multimedia projects across the curriculum or whole-class demonstrations of manipulatives in action, book pages, science models, and more. Connects via USB port to use with your computer or interactive whiteboard. No batteries required. Compatible with PC and Mac.
Height: 20''
Spécifications
- Résolution:3 MP
- Désignation:Interactive Projection Camera