Tungsten Carbide Cutters, Electron Microscopy Sciences
Tungsten Carbide Cutters, Electron Microscopy Sciences
Certificates
Tungsten Carbide Cutters, Electron Microscopy Sciences
Tungsten Carbide Cutters, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These cutters and pliers are designed for use in applications where strength, wear resistance, and hardness are a primary concern.
  • Magnetizable
  • Harden by heat treatment
  • ESD safe
Tungsten carbide and high precision cutters are made from carbon steel, with a maximum hardness of 68 HRC. They offer a moderate resistance to corrosion, and are ideal for cutting and bending single or multiple filars, and for lateral or internal cuts.
ESD epoxy-coated cutting tweezers have a poor resisance to corrosion. They are often used as tweezers in medical device manufacturing, and as a cutting tool in the electricity industry.
Product Family Options
High Precision Cutters
Informations sur le produit
  • Désignation
    Overall Length
    Tip Style
    Jaw Type
    Availability
    Price
  • High precision cutter, style ES5130M
    120 mm
    Micro oval
    Discontinued
    Supplier Number: -
  • High precision cutter, style ES5132M
    120 mm
    Micro oval
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $645.91
    $645.91
    Each
       $645.91
    Supplier Number: 9520
  • High precision cutter, style ES5140
    120 mm
    Small oval
    Discontinued
    Supplier Number: -
  • High precision cutter, style ES5141
    120 mm
    Small oval
    Discontinued
    Supplier Number: -
  • High precision cutter, style ES5142
    120 mm
    Small oval
    Discontinued
    Supplier Number: -
  • High precision cutter, style ES5152
    120 mm
    Large oval
    Discontinued
    Supplier Number: -
  • High precision cutter, style ES5152
    125 mm
    Large oval
    Discontinued
    Supplier Number: -
  • High precision cutter, style ES5150
    120 mm
    Large oval
    Discontinued
    Supplier Number: -
  • High precision cutter, style ES5162
    120 mm
    Extra large oval
    Discontinued
    Supplier Number: -
  • High precision cutter, style ES5160
    125 mm
    Extra large oval
    Discontinued
    Supplier Number: -
  • High precision cutter, style ES5161
    125 mm
    Extra large oval
    Discontinued
    Supplier Number: -
  • High precision cutter, style ES5340
    120 mm
    Small tapered
    Discontinued
    Supplier Number: -
  • High precision cutter, style ES5341
    120 mm
    Small tapered
    Discontinued
    Supplier Number: -
  • High precision cutter, style ES5342
    120 mm
    Small tapered
    Discontinued
    Supplier Number: -
  • High precision cutter, style ES5350
    120 mm
    Large tapered
    Discontinued
    Supplier Number: -
  • High precision cutter, style ES5351
    120 mm
    Large tapered
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $277.26
    $277.26
    Each
       $277.26
    Supplier Number: 9506
  • High precision cutter, style ES5352
    120 mm
    Large tapered
    Discontinued
    Supplier Number: -
  • High precision cutter, style ES5540
    120 mm
    Small tapered and relieved
    Discontinued
    Supplier Number: -
  • High precision cutter, style ES5541
    120 mm
    Small tapered and relieved
    Discontinued
    Supplier Number: -
  • High precision cutter, style ES5542
    120 mm
    Small tapered and relieved
    Discontinued
    Supplier Number: -
ESD Epoxy-Coated Cutting Tweezers
Informations sur le produit
  • Désignation
    Overall Length
    Tip Style
    Jaw Type
    Availability
    Price
  • Cutting tweezers, style 15AGWM
    115 mm
    Angled
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $406.69
    $406.69
    Each
       $406.69
    Supplier Number: 9500
  • Cutting tweezers, style 15ARWM
    116 mm
    Angled
    Discontinued
    Supplier Number: -
  • High precision cutting tweezer, style 152.S.0
    100 mm
    Angled blade
    Discontinued
    Supplier Number: -