Nitrile Examination Gloves, Textured, Electron Microscopy Sciences
À propos de cet article
Gloves are made from 100% synthetic nitrile that contains no latex. They offer superior textile strength and are puncture-resistant when compared to other latex or vinyl gloves of similar thickness.
- Textured finish provides a positive grip on wet and/or dry surfaces
- Excellent textile strength
- Puncture-resistant
- Latex-free
- Powder-free
These powder-free gloves will provide the user with superior comfort, excellent flexibility, and dexterity. The nitrile compound provides a broad range of hand protection against accidental chemical splashes. There are no natural rubber protein allergens as is present in natural latex rubber gloves.
4 Options Available Below
Product Details & Documents
Gloves are made from 100% synthetic nitrile that contains no latex. They offer superior textile strength and are puncture-resistant when compared to other latex or vinyl gloves of similar thickness.
- Textured finish provides a positive grip on wet and/or dry surfaces
- Excellent textile strength
- Puncture-resistant
- Latex-free
- Powder-free
These powder-free gloves will provide the user with superior comfort, excellent flexibility, and dexterity. The nitrile compound provides a broad range of hand protection against accidental chemical splashes. There are no natural rubber protein allergens as is present in natural latex rubber gloves.
Documents
Certificates
Recherche par numéro de lotSpecifications for Product Family
Spécifications
- Catalog No:
- 76458-564
- 76458-556
- 76458-558
- 76458-560
- 76458-562
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Textured
- Textured
- Textured
- Textured
- Textured
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
Spécifications
Product Family Options
Informations sur le produit
- Taille du gantGlove ColorAvailabilityPrice
- Supplier Number: 71116-90Spécifications:
More
Specifications
Cat. No.76458-564Disposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-freeFinishTexturedSterilityNon sterile - Supplier Number: 71116-91Spécifications:
More
Specifications
Cat. No.76458-556Disposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-freeFinishTexturedSterilityNon sterile - Supplier Number: 71116-92Spécifications:
More
Specifications
Cat. No.76458-558Disposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-freeFinishTexturedSterilityNon sterile - LBrownDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.76458-560Disposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-freeFinishTexturedSterilityNon sterile - Supplier Number: 71116-94Spécifications:
More
Specifications
Cat. No.76458-562Disposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-freeFinishTexturedSterilityNon sterile