Monarch® gDNA Elution Buffer, New England Biolabs
Catalog # CA76339-784
Supplier: New England Biolabs (NEB)
CAS Number:
Specifications
- Description:Monarch® gDNA elution buffer
- Size:34 ml
- Cat. no.:CA76339-784
Specifications
About this item
The Monarch® gDNA Elution Buffer a component of the Monarch® Genomic DNA Purification Kit (NEB #T3010), which can be used to purify up to 30 µg genomic DNA from a variety of sample types.
Monarch® gDNA Elution Buffer is supplied in 10 mM Tris-HCl, pH 9.0, 0.1 mM EDTA and is suitable for long term storage of gDNA. Nucleases are inactivated both by the inclusion of EDTA and the pH of the solution.