ContactUsLinkComponent
Monarch® gDNA Wash Buffer, New England Biolabs
Catalogue # CA76339-782
Numéro CAS:  
Monarch® gDNA Wash Buffer, New England Biolabs
Catalogue # CA76339-782
Supplier Number:  T3015L
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Monarch® gDNA wash buffer
  • Size:
    60 ml
  • Cat. no.:
    CA76339-782
Spécifications
À propos de cet article
The Monarch® gDNA Wash Buffer a component of the Monarch® Genomic DNA Purification Kit (NEB #T3010), which can be used to purify up to 30 µg genomic DNA from a variety of sample types.
Monarch® gDNA Wash Buffer is a component of the Monarch® Genomic DNA Purification Kit (NEB #T3010). It is an ethanol-based, low salt buffer that allows for complete removal of chaotropic salt and cell components in just 2 wash steps, enabling the purification of highly-pure gDNA.