ContactUsLinkComponent
Monarch® gDNA Blood Lysis Buffer, New England Biolabs
Monarch® gDNA Blood Lysis Buffer, New England Biolabs
Catalog # CA76339-778
CAS Number:  
Monarch® gDNA Blood Lysis Buffer, New England Biolabs
Catalog # CA76339-778
Supplier Number:  T3013L
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Monarch® gDNA blood lysis buffer
  • Size:
    20 ml
  • Cat. no.:
    CA76339-778
Specifications
About this item
The Monarch® gDNA Blood Lysis Buffer a component of the Monarch® Genomic DNA Purification Kit (NEB #T3010), which can be used to purify up to 30 µg genomic DNA from a variety of sample types.
The kit contains three unique lysis buffers, optimized to enable maximal yield and purity when preparing genomic DNA from a variety of sample types. The Blood Lysis Buffer contains a strong protective component against the high nuclease activity in blood samples and supports rapid degradation of hemoglobin and other protein components.
The Monarch® gDNA Blood Lysis Buffer is one of three lysis buffers available for purchase separately for additional convenience and to minimize waste.