ContactUsLinkComponent
Monarch® RNase A, New England Biolabs
Monarch® RNase A, New England Biolabs
Catalog # CA76339-788
CAS Number:  
Monarch® RNase A, New England Biolabs
Catalog # CA76339-788
Supplier Number:  T3018L
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    RNase A, 20 mg/ml concentration
  • Size:
    1 ml (2×0.5 ml)
  • Cat. no.:
    CA76339-788
Specifications
About this item
Monarch® RNase A is a component of the Monarch® Genomic DNA Purification Kit (NEB #T3010), which can be used to purify genomic DNA from a variety of sample types. 
Monarch® RNase A is a component of the Monarch® Genomic DNA Purification Kit (NEB #T3010), which can be used to purify genomic DNA from a variety of sample types. Monarch® RNase A degrades single-stranded RNA at C and U residues. This formulation is purified from cow pancreas, is free of contaminating protease or DNase activity, and contains no residual host DNA. It is used at 56 °C in the Monarch® Genomic DNA Purification Kit protocol but is active at any temperature between 20 and 65 °C. It is supplied as a 20 mg/ml solution in 50% glycerol, 50 mM Tris-Cl pH 7.4.
Treatment with RNase A is an optional step in the Monarch® Genomic DNA Purification Kit protocol for removal of any residual RNA. Co-purification of RNA during gDNA extractions is a common problem that leads to the overestimation of DNA yield. The low alcohol binding conditions employed in the kit optimizes binding of gDNA alone, so RNA co-purification is extremely low; however, an RNase A treatment step is included in each protocol as an option for the user.