Intelsius® BioTherm (2...8 °C) Sample Transport Shippers, DGP Intelsius
Fournisseur: DGP INTELSIUS LLC
Certificats
À propos de cet article
Intelsius® BioTherm shippers are suitable for shipping temperature critical samples requiring a stable fridge temperature of between 2 and 8 °C.
- Passive cooling
- Compliant labels
- Reusable cold pack technology
- Regulatory compliant packaging
The BioTherm range of high performance 2 to 8 °C solutions are fully compliant to IATA, ADR, and DOT (49 CFR) transport regulations. These systems are qualified to ensure sample integrity for up to 96 hours against 2 to 8 °C temperature exposures. Each system is provided with handles and the correct compliance labeling to enable careful handling throughout the regulatory compliant cold chain. Cold packs are included.
Frequently Bought Together