Regular Polyester Tip Swab with Plastic Applicator, Sterile
Catalog # 76452-298
Supplier: Hardy Diagnostics
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Sterility:Sterile
- Color:White
- Description:Polyester tip swab with 100 mm breakpoint
- Material:Polyester
- Cat. no.:76452-298
- Packaging:Peel pouch, individually wrapped
Specifications
About this item
Standard polyester tip swabs feature a plastic applicator shaft with a 100 mm breakpoint.
They can be used with most PCR platforms.
Packaging: These sterile swabs are supplied individually wrapped in a peel pouch.