Intelsius® Biotherm Packaging Systems are regulatory compliant to IATA, ADR, and 49 CFR (DOT) transport Regulations. These shippers utilize dry ice to maintain sample integrity during shipments from 24 to 240 hours.
- Regulatory compliant
- Dry ice shippers
- Compliant markings on boxes
- Variety of internal volumes
Intelsius® Biotherm shipping systems are pre-qualified to maintain sample integrity during shipments from 24 to 240 hours against demanding high-performance dry ice temperature (2 to 8 °C profiles and 15 to 25 °C profiles). The range is tested and qualified in conjunction with Intelsius® Specimen Transport Packaging.
The range is designed to optimize shipping volume and minimize freight costs. These insulated shipping systems have unique, sturdy carry handles to enable careful handling throughout the cold chain and a tongue and groove lid and base to enable quick and simple assembly. Pack-out instruction sheets are supplied to ensure correct assembly.